Imprimer
Ion selective electrode, half-cell, thiocyanate, Orion™
Fournisseur: Thermo Orion
9458BN
ORIO9458BNEA 2000 EUR
ORIO9458BN
Ion selective electrode, half-cell, thiocyanate, Orion™
Électrodes Électrodes ISE
This half-cell electrode is ideal for performing quick, accurate and economical measurements of thiocyanate ions in aqueous solutions.
  • Solid-state half-cell requires reference electrode
  • pH < 4 is recommended to avoid OH-interference
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR