Imprimer
pAAV-AC-GFP AAV Gene Expression Vector
Fournisseur: OriGene
PS100090
ORIGPS100090EA 1180 EUR
ORIGPS100090
pAAV-AC-GFP AAV Gene Expression Vector
Vecteurs, plasmides et librairies
AAV Gene Expression Vectors with C-terminal tGFP tag.
  • OriGene's TrueORF clones can be shuttled into this vector via PrecisionShuttle, a simple cut-and-ligation process
  • The ORF cloned in this vector can be expressed containing c-terminal tGFP
  • BGI_F Primer can be used to sequence the target sequence after cloning
  • AC_GFP_R Primer can be used to sequence the target sequence after cloning
E. coli Selection: Ampicillin (100 µg/ml)
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR