Imprimer
Vial crusher, national
Fournisseur: THERMO NATIONAL SCIENTIFIC
C4040-VC
NSCAC4040-VCEA 12200 EUR
NSCAC4040-VC
Vial crusher, national
Ouvre-ampoules
Breaks and separates liquid from all 1 ml to 100 ml vials and ampoules.
  • Capable of processing approximately 5000 of 2 ml vials per hour
  • Reduces overall waste disposal costs by reducing the total volume of waste
Safe use in a fume hood or connect to a vacuum for limited exposure to flammable or toxic vapours. Environmentally responsible waste handling, crushed glass is automatically separated from solvent for separate disposal.
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR