Imprimer
Spoons, stainless steel
Fournisseur: NICKEL ELECTRO
8040
NICK8040EA 11.7 EUR
NICK8040
Spoons, stainless steel
Cuillères
These laboratory spoons are made of corrosion-resistant 304 stainless steel.
  • Durable surface is easy to clean
  • Non-magnetic
  • Bright finish
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR