Imprimer
PIG® high-visibility yellow mat pads and rolls
Fournisseur: NEW PIG
MAT604 MAT630 MAT605
NEWPMAT604EA 127 EUR
NEWPMAT604 NEWPMAT630 NEWPMAT605
PIG® high-visibility yellow mat pads and rolls
Matériaux absorbants
Hi-viz spill absorbents alert workers to slip hazards while cleaning up leaks.
  • High-visibility yellow colour is easier to see in poorly lit areas to minimise tripping hazards; alerts workers to potential dangers and work in progress
  • Eight layers of 100% polypropylene are thermally bonded to make PIG Mat the strongest mat on the market; won't rip, tear or fray even when saturated
  • Exclusive dimple pattern speeds wicking of liquid throughout mat for faster, easier cleanup
  • Highly absorbent, fine-fibre construction won't leave liquids or fibre residue behind
More Product Information
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR