Imprimer
PIG® Sticky Steps® mat
Fournisseur: NEW PIG
MAT566-BL
NEWPMAT566-BLEA 112 EUR
NEWPMAT566-BL
PIG® Sticky Steps® mat
Matériaux absorbants
Multi-sheet floor mat grabs grime from shoes and wheels to keep dirt out of offices, cleanrooms and other areas where clean floors are vital.
  • High-tack disposable mat pulls dirt and debris from shoes and wheels to prevent tracking into offices
  • Used sheets tear off easily and individually
  • Numbered pull-tab system shows when you're running low
  • Low-profile pack adheres to floor with easy-to-use backing
  • Each 4 mil polyethylene sheet is nonallergenic, nonodourous and nondrying
  • Walking surface: Sticky top
Weight: 6,35 kg
Thickness: 100 micron
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR