Imprimer
Cryoboxes
Fournisseur: NEOLAB
7-8040
NEOL7-8040EA 139 EUR
NEOL7-8040
Cryoboxes
Boîtes Boîtes pour cryotubes
Boxes are made of polypropylene for the storage of light-sensitive samples. 100 places 12,5×12,5 mm for 0,5, 1,5, 2,0 ml microcentrifuge and cryo tubes.
  • Designed for use in deep freezers at temperature of –90 to +120 °C
  • Alphanumerical index on the lid for identifying contents
  • Stackable
  • For max. 53 mm tube height
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR