Imprimer
Muffle furnaces with lift up door, Tmax 1300 °C and controller C550, LT 5/13 - LT 15/13
Fournisseur: NABERTHERM
L-055H3ANO L-055H3ANOG L-055H3ANOGT L-055H3ANOGTZ L-055H3ANOGZ L-055H3ANOT L-055H3ANOTZ L-055H3ANOZ
NABEL-055H3ANOEA 5860 EUR
NABEL-055H3ANO NABEL-055H3ANOG NABEL-055H3ANOGT NABEL-055H3ANOGTZ NABEL-055H3ANOGZ NABEL-055H3ANOT NABEL-055H3ANOTZ NABEL-055H3ANOZ
Muffle furnaces with lift up door, Tmax 1300 °C and controller C550, LT 5/13 - LT 15/13
Étuves Fours
These furnaces feature heating elements on supporting tubes that radiate freely into the chamber, ensuring particularly short heating times. With the robust lightweight refractory brick insulation, they can reach a max. working temperature of 1300 °C.
  • Heating from two sides
  • Multi-layer insulation with robust lightweight refractory bricks in the furnace chamber
  • Textured stainless steel housing
  • Dual shell housing for low external temperatures and high stability
  • Lift up door (LT) with hot surface facing away from the operator
  • Exhaust air outlet on rear wall
More Product Information
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR