Imprimer

Potassium iodure 20% aqueous solution

Fournisseur: MARKET SOURCE PART PROCESS

Synonyms: Dipotassium diiodide, Kali iodide

80012284.1000
MSPP80012284.1000EA 0 EUR
MSPP80012284.1000
Potassium iodure 20% aqueous solution
Potassium iodure
Formule: Kl
MW: 166 g/mol
Point d'ébullition: 1330
Point de fusion: 686
Température de stockage: Température ambiante
Numéro MDL: MFCD00011405
Numéro CAS: 7681-11-0
EINECS: 231-659-4
Merck Index: 13,07727

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR