Imprimer
Universal drum tops
Fournisseur: asecos
9953
MSGA9953EA 127 EUR
MSGA9953
Universal drum tops
Matériaux absorbants
Universal absorbent mat for drum tops for safe absorption of oil, water, cooling liquids and non aggressive chemicals.
  • Suitable for 200 litre drums
  • Optimal protection while dispensing ensuring drum top and floor stay clean
  • Absorbent capacity up to 16 times its own weight
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR