Imprimer
Drum taps, stainless steel
Fournisseur: asecos
2795 2797
MSGA2795EA 239 EUR
MSGA2795 MSGA2797
Drum taps, stainless steel
Robinets et éviers Robinets
These drum taps are made of stainless steel.
  • Adjustable removal position
  • Self closing
  • Flame guard
  • PTFE seal
Informations de commande: MSGA2797 includes a trailer fitting for decanting vessels.
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR