Imprimer

Shelving systems for hazardous substances, RWW

Fournisseur: asecos
27829 27830 27831 27832 27833 27834 27835 27836
MSGA27829EA 818 EUR
MSGA27829 MSGA27830 MSGA27831 MSGA27832 MSGA27833 MSGA27834 MSGA27835 MSGA27836
Shelving systems for hazardous substances, RWW
Mobilier Étagères
Sturdy construction racks with four galvanized steel sumps shelves as storage levels and galvanized rack frames. Ideal for safe and approved storage of water pollutants.

  • Storage levels in 25 mm increments, height-adjustable
  • Load capacity per storage shelf: 200 kg (evenly distributed)
  • Rack system can be extended individually

Certifications: TÜV approved.

Information de livraison: Flat-pack delivery with assembly instructions.
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR