Imprimer
Autoclave test packs, Bowie and Dick type, Comply™
Fournisseur: 3M MEDICAL
M1301
MMMEM1301EA 387 EUR
MMMEM1301
Autoclave test packs, Bowie and Dick type, Comply™
Indicateurs de stérilisation
With unique colour chemistry, the 3M™ Comply™ Disposable Bowie and Dick Type Autoclave Test Pack has been designed for single use in porous load sterilizers for daily Bowie and Dick tests.
  • Easy to detect failures according to ISO 11140 Part 1
  • Easy to interpret fail or pass
  • Easy to locate and retrieve test sheet
  • Easy to open "matchbox" system
  • Paper indicator sheet
Bowie-Dick Test Packs are used to monitor the performance of vacuum-assisted pre-vac steam sterilizers; test packs will reproducibly detect residual air in the sterilizer chamber due to leaks,More Product Information
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR