Imprimer
Faceshield Holder, V4
Fournisseur: 3M
V46
MMMAV46EA 0 EUR
MMMAV46
Faceshield Holder, V4
Écrans de protection
Face shield holder for V4-Series face shields. This face shield holder with short peak is designed for use with 3M™ V4-Series face shields.
  • Replacement face shield frame
  • Short peak design
  • For use with 3M™ V4 Series face shields alongside 3M™ linkage kit V412
Ensure optimum comfort and protection from a hard hat with 3M™ Faceshield Holder, V46. This faceshield holder has a short peak, and when used with the 3M™ Linkage Kit V412 can quickly and easily attach a faceshield to a 3M™ Hard Hat.
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR