Imprimer
Reporter gene assays, SpectraMax® DuoLuc™ reporter assay kit
Fournisseur: Molecular Devices
R8361
MLDVR8361EA 369 EUR
MLDVR8361
Reporter gene assays, SpectraMax® DuoLuc™ reporter assay kit
Tests Tests cellulaires
SpectraMax® DuoLuc™ reporter assay kit offers sensitive detection of dual luciferase reporter gene assays, which are important tools for monitoring gene expression and other cellular events.
  • Enables highly sensitive quantitation of both firefly and Renilla luciferases in mammalian cells
  • Flash-type luminescence with excellent sensitivity
  • Expression measurement in as few as ten cells per well
  • Improved accuracy of results by normalising reporter expression to a control
  • Optimised for SpectraMax® i3x, iD3, or L microplate readers with injectors
  • Simplified data acquisition and analysis with preconfigured protocol in SoftMax® Pro Software
Commander maintenant
SPÉCIFICATIONS
Environmentally Preferable
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR