Imprimer
SNAP i.d. 2.0 Protein Detection Systems Mini and Midi
Fournisseur: Merck
TISNAP2MM
MILFTISNAP2MMEA 0 EUR
MILFTISNAP2MM
SNAP i.d. 2.0 Protein Detection Systems Mini and Midi
Analyseurs Analyseur de protéines
Developed to meet the needs of our Western blotting customers, the SNAP i.d. 2.0 system produces blots of a very high quality. Unique vacuum-driven technology and a built-in flow distributor actively drive reagents through the membrane.
Informations de commande: This system includes the SNAP i.d. 2.0™ base, a mini blot holding frame,a midi blot holding frame, two antibody collection trays, vacuum tubing, rolling pad and blot roller, two wetting trays and a quick start guide.
Commander maintenant
SPÉCIFICATIONS
Environmentally Preferable
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR