Imprimer
Millicell® EZ Slide Holder
Fournisseur: Merck
PEZXMSH01
MILFPEZXMSH01EA 195 EUR
MILFPEZXMSH01
Millicell® EZ Slide Holder
Porte-lames
Microscope slide holder for 4 and 8 well units, holds two slides.
  • Shipped in ambient temperature
Techniques: cell based assay: suitable cell culture; mammalian: suitable immunocytochemistry: suitable
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR