Imprimer
Millipore® Immobilon®-PSQ NOW Bundle Hydrophobic PVDF Transfer Membrane
Fournisseur: Merck
NWKITSQ
MILFNWKITSQEA 1030 EUR
MILFNWKITSQ
Millipore® Immobilon®-PSQ NOW Bundle Hydrophobic PVDF Transfer Membrane
Membranes de transfert
PVDF transfer membrane hydrophobic for western blotting using chemiluminescence, chromogenic or radioactive detection methods.
  • Western blotting
  • Dot blot
  • White/plain filter
  • Electrostatic, hydrophobic
  • 0,2 µm filter pore size
  • Comes in an 8,5 cm × 10 m roll
The Immobilon NOW system offers 1-cut convenience for standard mini-blots (7×8,5 cm) and midi-blots (8,5×13,5 cm).
Compatible stainn: Coomassie brilliant blue, Amido black, India ink, Ponceau-S red, Colloidal gold, CPTS, Toluidine blue, Transillumination and Sypro® ruby.
New Product
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR