Imprimer
Membrane filter, silver
Fournisseur: Merck
AG4502550
MILFAG4502550EA 806 EUR
MILFAG4502550
Membrane filter, silver
Filtres Filtres à membrane
These silver membrane filter are suitable for a variety of applications and are commonly used for air monitoring and analysis of particulate contaminants. They are specified for collection and detection of particles such as silica, carbon black, coal tar products and coke oven emissions.
  • Pure silver filters for particulate monitoring
  • Highly resistant to thermal stress and aggressive chemicals
  • Low background for sensitive X-ray diffraction analysis
Used for air monitoring: asbestos, lead sulphide, crystalline and amorphous silica.
Commander maintenant
SPÉCIFICATIONS
Environmentally Preferable
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR