Imprimer
Construction tape, polyethylene
Fournisseur: MICRONOVA
CTP-2WH
MICRCTP-2WHEA 24.4 EUR
MICRCTP-2WH
Construction tape, polyethylene
Rubans adhésifs
This high-tack tape has rubber adhesive that easily bonds to plastic, concrete, and stainless steel.
  • Excellent conformity
  • Serrated edge for easy tearing by hand
  • Polyethylene fabrication
Tape conforms well and does not have the same 'dead stretch' limitations common in other polyethylene tapes. Unique serrated edge allows an easy tear at any length, for environments where blades and dispensers are not practical.
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR