Imprimer
Bucket Liners
Fournisseur: MICRONOVA
BKL1-1818IR BKL-2430IR BKL-2424IR BKL7-3119 BKL99-3521IR BKL99-3521
MICRBKL1-1818IREA 444 EUR
MICRBKL1-1818IR MICRBKL-2430IR MICRBKL-2424IR MICRBKL7-3119 MICRBKL99-3521IR MICRBKL99-3521
Bucket Liners
Sachets Sacs à déchets
The unique LDPE/nylon laminate is a good barrier material for transporting raw materials or to line buckets containing disinfectants or other cleaning solutions.
  • Laminate is 75% LDPE, 25% nylon
  • Excellent for disinfectants and cleaning solutions
  • Ideal for product parts transport
Designed for cleanrooms and controlled environments. Available as sterile and non-irradiated.
Conditionnement: Individually double packed, packed in a case of 24/50/200.
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR