Imprimer
Masterflex® B/T® PerfectPosition® Pump Tubing, Peroxide-Cured Silicone, Avantor®
Fournisseur: Avantor Fluid Handling
96400-90
MFLX96400-91EA 726 EUR
MFLX96400-91 MFLX96400-90
Masterflex® B/T® PerfectPosition® Pump Tubing, Peroxide-Cured Silicone, Avantor®
Tuyaux Tubes pour pompe péristaltique
Ensure optimum performance from your Masterflex® pump.
  • Excellent biocompatibility
  • No additives, plasticizers or DOP; odorless and nontoxic, fungus-resistant
  • No taste imparted to transported fluids
  • Extremely good at low temperatures
  • Weather, ozone, corona, and radiation resistant
  • Minimal tendency to take a set
  • PerfectPosition™ tubing retention marks indicate the exact placement of tubing in the pump head to provide the best performance and life of the tubing
  • Custom extruded to fit 77110- and 77111-series Masterflex® B/T® pumps and pump heads
  • Engineered for
More Product Information
Plus About VWR