Imprimer
Dow Corning® Pharma-65 Transfer Tubing
Fournisseur: Avantor Fluid Handling
96107-09
MFLX96107-01EA 225 EUR
MFLX96107-01 MFLX96107-04 MFLX96107-05 MFLX96107-11 MFLX96107-00 MFLX96107-02 MFLX96107-09
Dow Corning® Pharma-65 Transfer Tubing
Tuyaux Transfer Tubing
Platinum-cured silicone tubing with a very low extractable profile.
  • Global market leader in range of formulations for most extensive chemical compatibility
Applications: Pharmaceutical, biotechnology, and ultra-pure fluid processing.
Characteristics: Platinum-cured silicone tubing with a very low extractable profile; it contains no phthalates, plasticizers, latex, or other organic additives. Remains flexible and resilient for long service life. Dow Corning maintains complete traceability from silicon to silicone tubing. Durometer shore A rating of 62.
More Product Information
Plus About VWR