Imprimer
Masterflex® Self-Priming Petroleum Centrifugal Pumps, Avantor®
Fournisseur: Avantor Fluid Handling
79735-11
MFLX79735-11EA 2070 EUR
MFLX79735-11
Masterflex® Self-Priming Petroleum Centrifugal Pumps, Avantor®
Pompe Centrifugal Pumps
Compatible with three distinct fuel classifications.
  • Models for the following fluid groups:- Gasoline, kerosene, avgas, and jet fuel- Biodiesel, fuel oil, and diesel- Ethanol and E85 blend
  • Available in both self-priming and standard end suction configurations
  • Custom configurations are a phone call away to our Application Specialists
These close-coupled pumps have been specifically engineered for compatibility with clean, nonabrasive petroleum products. Transfer and delivery of various fuels such as gasoline, ethanol, biodiesel, and fuel oils are justMore Product Information
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR