Imprimer
Edwards Vacuum Solenoid Right-Angle Valves
Fournisseur: Avantor Fluid Handling
79302-78
MFLX79302-78EA 1820 EUR
MFLX79302-78
Edwards Vacuum Solenoid Right-Angle Valves
Vannes Solenoid Valves
Designed to maintain vacuum levels in your systems.
  • Easily control vacuum levels with these solenoid valves
Compact and lightweight valves offer a pressure range from 8 x 10⁻¹⁰ to 150 Torr.
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR