Imprimer
Almatec Air-driven Postive Displacement Pumps
Fournisseur: Avantor Fluid Handling
75980-04 75980-16
MFLX75980-04EA 3150 EUR
MFLX75980-04 MFLX75980-16
Almatec Air-driven Postive Displacement Pumps
Pompe Pompes pneumatiques
German precision engineered AODD pumps to get your application up and flowing.
  • Metal-free air-operated double diaphragm pumps
The heart of the AODD is the air-valve and thoughtful German engineering went into the development of the precision control PERSWING P® air valve and its Air Distribution System (ADS). Resulting in a metal-free, pneumatically pilot-operated PERSWING P® (ADS) that ensures accurate reversal of the main piston and offers low noise levels, minimal maintenance and lube-free operation (due to two moving parts).
Almatec plastic pumpsMore Product Information
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR