Imprimer
Abaque™ CSM Peristaltic Pump Replacement Hoses
Fournisseur: Avantor Fluid Handling
MFLX75810-36EA 646 EUR
MFLX75810-36
Abaque™ CSM Peristaltic Pump Replacement Hoses
Tuyaux Tubes pour pompe péristaltique
High resistance to oxidants, as well as to concentrated bases and acids.
  • All listed hoses in this ad are made from CSM.
  • The hose IDs are: 10, 15, 20, 25, 32, 40, 50, 65, 80, 100 mm. Each liquid end is designed to accept one particular hose ID version.
The hose of a Peristaltic Hose pump is the only component that comes in contact with the pumped medium. Neptune engineers hoses the are manufactured from high quality compounded rubber, reinforced by layers of nylon. Once constructed, the hose is then machined with precision for efficiency, accuracy,More Product Information
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR