Imprimer
Masterflex® Analog Gear Pump Systems, Avantor®
Fournisseur: Avantor Fluid Handling
74013-52
MFLX74013-02EA 2910 EUR
MFLX74013-02 MFLX74013-07 MFLX74013-12 MFLX74013-22 MFLX74013-32 MFLX74013-42 MFLX74013-47 MFLX74013-57 MFLX74013-72 MFLX74013-17 MFLX74013-27 MFLX74013-37 MFLX74013-52 MFLX74013-77 MFLX74013-77-EU
Masterflex® Analog Gear Pump Systems, Avantor®
Pompe Gear Pumps
Simple variable speed control handles many fluid handling applications.
  • 100:1 speed-control range controls flow over a wide range
  • Simple operation with single-turn speed control
  • Separate power switch enables you to maintain speed setting when turning drive on/off; front-panel LED indicates power on
  • Remote control of start/stop via DB9 connector on back of drive
Compact, cost-effective drive offers simple variable speed control for many fluid handling applications from general transfer to filtration to chemical injection. Conveniently coupled with an A-More Product Information
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR