Imprimer
Masterflex® Luer Winged Fitting, Avantor®
Fournisseur: Avantor Fluid Handling
MFLX40325-00EA 12.8 EUR
MFLX40325-00 MFLX40325-01 MFLX40325-02 MFLX40325-03 MFLX40325-07 MFLX40325-06 MFLX40325-05 MFLX40325-04
Masterflex® Luer Winged Fitting, Avantor®
Raccords et connecteurs
Easily transition to a luer fitting.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Determine which connector type is needed for your application by contacting free technical application assistance
These fittings are manufactured in an ISO Class 7 cleanroom at an ISO 9001 facility; utilizing an ISO 13485 quality system. There is full lot traceability and tight dimensional tolerances forMore Product Information
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR