Imprimer
Masterflex® I/P® Variable-Speed Air-Powered Drive, Avantor®
Fournisseur: Avantor Fluid Handling
MFLX07589-30EA 2880 EUR
MFLX07589-30
Masterflex® I/P® Variable-Speed Air-Powered Drive, Avantor®
Pompe Pump Drives Peristaltic Pump Drives
Pump safely in hazardous environments.
  • Flow rate 0.6 to 13 LPM I/P tubing—flow rate depends on drive rpm, pump head, and tubing size
  • Easily adjust motor speed by using included regulator
  • Effortlessly connect to your existing air supply or compressor
  • The 3/4-hp air-powered motor starts smooth, runs cool, and requires little maintenance
  • ATEX Zone 2 approved; rated to EEXII 3 G c IIC T6; NEC Class 1, Division 2, Groups A, B, C, D, T6
Use this variable-speed air-powered drive when electrical power may be unsafe or impractical. For safe operation,More Product Information
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR