Imprimer
Masterflex® L/S® Standard Pump Head Thrust Washers, Avantor®
Fournisseur: Avantor Fluid Handling
MFLX07021-04EA 37.6 EUR
MFLX07021-04
Masterflex® L/S® Standard Pump Head Thrust Washers, Avantor®
Pompe Pump Heads Peristaltic Pump Heads
Get the most out of your Masterflex® L/S® Standard Pump Head with these replacement parts.
  • Suitable for use with Masterflex® L/S® Standard pump heads
Replacement parts to keep your pump head operating even longer. Thrust washers fit on either side of the rotor assembly of L/S standard pump heads.
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR