Imprimer
In-Line Flow Indicators for Liquids and Gases
Fournisseur: Avantor Fluid Handling
06297-00
MFLX06297-00EA 70.8 EUR
MFLX06297-00
In-Line Flow Indicators for Liquids and Gases
Flow Indicators
An inexpensive way to visually monitor flow of liquids and gases.
  • Visually confirm flow with clear window
Indicators give visual confirmation of flow through a clear polystyrene housing. All models can be used with noncorrosive liquids and gases. Connect tubing to indicators via hose barb connections; model 06297-05 has straight ends. All models are pressure rated to 10 psig.
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR