Imprimer
TableWorx Performance Work Table, 304 Stainless Steel Work Surface
Fournisseur: Metro International
TWS3072SU304S
METOTWS3072SU304SEA 2784 EUR
METOTWS3072SU304S
TableWorx Performance Work Table, 304 Stainless Steel Work Surface
Mobilier Tables
TableWorx is a truly modular platform of performance work tables designed to allow you to customize your perfect solution.
  • Type 304 stainless steel work surface reinforced with 16-gauge hat channels
  • The sound deadened work surface features 90-degree turned down edges on all four sides
  • Available with an optional 51 mm (2") stainless steel backsplash, which is formed into the work surface
  • It features type 304 stainless steel legs and leg mounts
  • Silver colour
Stationary work table legs include stainless steel levelling bullet feet with 29 mm (1,125")More Product Information
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR