Imprimer

L(+)-Glutamine solution_SC 200 mM (100x) water + 8.5 g/L NaCl Cell Culture, Corning®

Fournisseur: Corning

Synonyms: Gln, L-Glutamine, H-Gln-OH, (S)-(+)-Glutamine, (S)-2,5-Diamino-5-oxopentanoic acid, Levoglutamide, Q

25-005-CV
MDTC25-005-CVEA 577.22 EUR
MDTC25-005-CV
L(+)-Glutamine solution_SC 200 mM (100x) water + 8.5 g/L NaCl Cell Culture, Corning®
L(+)-Glutamine

L-glutamine is an essential amino acid and a key component of culture media, serving as a major energy source for propagating cells. It is very stable as a dry powder and as a frozen solution but degrades rapidly in liquid media or stock solutions, producing toxic compounds. Optimal cell performance usually requires supplementation of the media with L-glutamine prior to use. This formulation is prepared in cell culture grade water.


Attention: Storage -25°C to -15°C

Formule: C₅H₁₀N₂O₃
MW: 146.15 g/mol
Point d'ébullition: 355
Numéro MDL: MFCD00008044
Numéro CAS: 56-85-9
Merck Index: 13,04484

Commander maintenant

Résultats des tests de spécification

pH 5.5 ± 0.5
Osmolality 450 ± 50 mOsm/kg H2O
Endotoxin < 0.25 EU/ml @ 1 :50
Mycoplasma Tested Negative
Sterility Pass
Cell Growth Pass


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR