Imprimer
Portable water sampler, P6
Fournisseur: MAXX
2070001
MAXA2070001EA 3370 EUR
MAXA2070001
Portable water sampler, P6
Échantillonneurs Échantillonneurs d'eau
This portable water sampler is a unique combination of design and technology as well as an unrivaled measuring device for volume determination.
  • Highly accurate sample volume with LED status display
  • Clear operating structure and simple programming
  • LAN/W-LAN/Web communication
  • Modern and ergonomic design
  • Easy cleaning
Long battery run-time thanks to 'sleep mode'. Integrated pump replacement tube at device versions with peristaltic pump.
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR