Imprimer
Urine test strips, Medi-Test
Fournisseur: MACHEREY-NAGEL
93001 93005 93006 93007 93009 93015 93021 93022 93023 93024 93027 93028 93029 93032 93035 93036 93037 93055 93058 93060 93067 93068
MANA93001EA 7.7 EUR
MANA93001 MANA93005 MANA93006 MANA93007 MANA93009 MANA93015 MANA93021 MANA93022 MANA93023 MANA93024 MANA93027 MANA93028 MANA93029 MANA93032 MANA93035 MANA93036 MANA93037 MANA93055 MANA93058 MANA93060 MANA93067 MANA93068
Urine test strips, Medi-Test
Systèmes de diagnostic clinique Réactifs et Kits de diagnostic clinique
Due to the high resistance towards interferences by ascorbic acid, a second testing for sensitive parameters such as blood or glucose is unnecessary in most cases.
  • User-friendly for quick results
  • Small amounts of urine can be checked
  • Optimised flexible shape of the strips to allow the examination of very small amounts of urine
Indispensable advantage especially in the field of pediatrics.
Commander maintenant
SPÉCIFICATIONS
Environmentally Preferable
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR