Imprimer

Universal pathogen DNA kit, Mag-Bind®

Fournisseur: OMEGA BIO-TEK

M4029-00 M4029-01
M4029-00EA 413 EUR
M4029-00 M4029-01
Universal pathogen DNA kit, Mag-Bind®
Réactifs pour acides nucléiques Nucleic Acid Purification Kits and Reagents

The Mag-Bind® Universal Pathogen DNA 96 Kit allows rapid and reliable isolation of high-quality pathogen and host genomic DNA from tissue samples. Up to a 30 mg tissue sample is recommended and can be processed in less than 60 minutes. The system allows for automation after sample lysis via Hamilton STAR™, Thermo KingFisher™ Flex, Applied Biosystems® MagMAX™, Qiagen Biosprint, and other liquid handling instruments.


The system combines the Mag-Bind® technology with RBB Buffer to eliminate PCR inhibiting compounds within the samples. Purified DNA is suitable for PCR, restriction digestion, and hybridisation applications.There are no organic extractions, thus reducing plastic waste and hands-on time and multiple samples can be processed in parallel.

Commander maintenant

SPÉCIFICATIONS

Désignation Mag-Bind® Universal Pathogen DNA Kit
Environmentally Preferable
Application
Target molecule DNA
Refine target molecule Viral DNA,Bacterial DNA
Sample type Stool,Biofluids (serum, plasma, saliva),Cultured cells,Human/animal cells and tissues
Method / Format Magnetic beads
Prep size Scalable
Taille de l'échantillon Variable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR