Imprimer
VWR® Antibody signal enhancer
Fournisseur: VWR Chemicals
M336-1LEA 0 EUR
M336-1L
VWR® Antibody signal enhancer
Systèmes de liaison et de marquage des protéines
Antibody Signal Enhancer is a Western blotting antibody diluent that increases intensity of protein detection up to 5-fold compared to conventional blotting.
  • Protein-free solution eliminates cross-reaction with antibodies
  • Reduces antigen and reagent use without the need for additional incubation steps
  • Formulated for use with HRP- and biotin-conjugated antibodies and works with both PVDF and nitrocellulose blots
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR