Imprimer
Stirring shafts
Fournisseur: Lenz Laborglas GmbH & CO.KG
06300510 06300516
LENZ06300510EA 36.9 EUR
LENZ06300510 LENZ06300516
Stirring shafts
Agitateurs Axes d'agitation
Stirring shafts made of precision calibrated glass tube.
  • Ground and polished bearing surface
  • Interchangeable
  • Made of DURAN® tubing
Commander maintenant
SPÉCIFICATIONS
Environmentally Preferable
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR