Imprimer
Storage cabinets for acid, Protector®
Fournisseur: LABCONCO
9901300
LBCP9901300EA 0 EUR
LBCP9901300
Storage cabinets for acid, Protector®
Mobilier Armoires de stockage
These cabinets support fume hoods and safely store and vent acids and also other corrosive liquids.
  • Manual closing door
  • Includes vacuum formed PVC liner tray
  • White exterior laboratory casework
  • Epoxy coated steel construction
Include one 20,3 cm (8") filler panel to increase cabinet’s overall depth from 55,9 cm (22") to 76,2 cm (30"), for installation flush to wall.
Informations sur les accessoires: Accessory interior shelf kits include powder-coated steel shelf, acid-resistant PVC tray and mounting hardware. The accessory vent kit is used forMore Product Information
Commander maintenant
SPÉCIFICATIONS
Environmentally Preferable
Learn more
About VWR
Plus About VWR