Imprimer
Ductless ventilation systems, Protector®XVS™
Fournisseur: LABCONCO
4863030 4864010 4864030
LBCP4863030EA 0 EUR
LBCP4863030 LBCP4864010 LBCP4864030
Ductless ventilation systems, Protector®XVS™
Hottes et coffrets Hottes filtrantes
Ductless ventilation systems with glass tops and sides and low profiles are well suited for educational settings. Clean-Sweep air foil and zone-perforated rear baffle maximise containment.
These ventilation stations can be used as light duty fume hoods, science classroom work stations, solvent cleaning work bays, weighing stations or for pathology and cytology.
Protector XVS ventilation stations can be ducted to the outside or connected to the FilterMate portable exhauster for HEPA and/or carbon filtered exhaust back into the laboratory.
Certifications: Conforms to ANSI Z9.5,More Product Information
Commander maintenant
SPÉCIFICATIONS
Environmentally Preferable
Learn more
About VWR
Plus About VWR