Imprimer

PuriCare® Procedure Stations with 12" Sash

Fournisseur: LABCONCO
312421031
LBCP312421031EA 0 EUR
LBCP312421031
PuriCare® Procedure Stations with 12" Sash
Hottes et coffrets Hottes pour la recherche animale
PuriCare Procedure Stations with a large sash opening create a particulate-free environment for small animal research activities such as cage changing, weighing, anesthesia delivery and surgical procedures.

  • Product and personnel protection
  • Large 12" sash opening
  • Erogonomic design

PuriCare Procedure Stations are Class II A2 biosafety cabinets that excel in safety, performance, and comfort. Their ergonomic design with the Inclination™ Technology Suite which includes the following: MyLogic™ operating system with eye-level color display, 10° angled sash,More Product Information
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR