Imprimer
Cuves pour Olli-C-Analyzer®
Fournisseur: KARTELL
2618
KART2618EA 311 EUR
KART2618
Cuves pour Olli-C-Analyzer®
Cuvettes Cuves de spécialités
These cylindrical cells are specifically used in conjunction with Olli-C-Analyzer.
  • Anti-shock polystyrene-moulding design
  • Highly resistant and dust-proof technology
  • Quality optical material suitable for IVD applications
Conditionnement: In polystyrene dust proof boxes of 192 pieces.
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR