Imprimer
Sterile Pipette Filter Tips in Rack
Fournisseur: KARTELL
0963500
KART0963500EA 360 EUR
KART0963500
Sterile Pipette Filter Tips in Rack
Pointes de pipettes
These tips are made of high quality, 100% pure, virgin PP. Made in the highest quality moulds, these tips can be used whenever the work demands a higher standard.
  • Sterilised by gamma radiation
  • Designed for use in molecular biology, cell cultures, bacteriology, virology, immunology and radioimmunology
  • Hydrophobic pe filters act as a barrier against aerosol contamination, preventing cross-contamination of samples and micropipettes
  • Tip orifices are sized to completely empty the tip in the event of over-aspiration
Made of PP and including a filter, theseMore Product Information
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR