Imprimer
Serological Pipettes Holder
Fournisseur: KARTELL
0111500
KART0111500EA 497 EUR
KART0111500
Serological Pipettes Holder
Portoirs pour pipettes
Bench pipette holder, made of particularly clear polymethylmethacrylate (PMMA), has high level finishes and its content can be seen from any angle.
  • Complete with four tilted support shelves and white top
The four separate compartments are ideal for housing plastic or glass serologic pipettes of various capacities (from 1 to 50 ml). Do not shine ultraviolet light on it.
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR