Imprimer

Colorant de Jenner, certified_Biological Stain Commission, Sigma-Aldrich®

Fournisseur: SIGMA ALDRICH MICROSCOPY

Synonyms: Eosin−Methylene Blue

J1875-25G
J1875-25GEA 212 EUR
J1875-25G
Colorant de Jenner, certified_Biological Stain Commission, Sigma-Aldrich®
Colorant de Jenner

Jenner's stain,certified by the Biological Stain Commission (Jenner Stain; Eosin-Methylene Blue)

Température de stockage: Température ambiante
Numéro MDL: MFCD00081733
Numéro CAS: 62851-42-7

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR