Imprimer
Colony Counter, Scan 50 Pro
Fournisseur: INTERSCIENCE
435055
INSC435055EA 1550 EUR
INSC435055
Colony Counter, Scan 50 Pro
Compteurs de colonies
Scan 50 pro is a manual colony counter with handpad and multiple ergonomic supports for improved posture, vision and hearing.
  • Compatible with all types of media
  • Contrasting colony lighting: Dark field 2
  • Touch surface with adjustable sensitivity
  • Ergonomics redesigned
  • Ninedifferent sounds and headphones jack to work without disturbing colleagues
  • Light signal for each counted colony
  • Optimal working posture, inclined reading surface with magnifying lens (optional), hand rest and HandPad
  • Ambidextrous - Duplicated count buttons on both sides
  • Easy to clean and
More Product Information
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR