Imprimer
Laboratory reactor systems, LR-2.ST the High Performer
Fournisseur: IKA
20013291
IKAA20013291EA 19010 EUR
IKAA20013291
Laboratory reactor systems, LR-2.ST the High Performer
Systèmes de réacteurs de laboratoire
Modular laboratory reactor for the optimisation and reproduction of various chemical reactions, mixing and homogenisation processes on a lab scale. The system is characterised by the agitator mounting, which allows a safe transfer of the higher motor torque.
  • Suitable for vacuum operation
  • Solvent- and temperature-resistant perfluoro elastomer (FFPM) seals come into contact with the sample
  • Infinitely variable speed
  • Torque trend display for measuring changes in viscosity
  • Microprocessor-controlled speed regulation, enables steady speed, also under load
More Product Information
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR