Imprimer
ROT.ROTATEMP SBS me-vi Viscometer Package
Fournisseur: IKA
10013993
IKAA10013993EA 7376 EUR
IKAA10013993
ROT.ROTATEMP SBS me-vi Viscometer Package
Viscosimètres Viscosimètres rotatifs
The ROTATEMP SBS me-vi package is the ideal solution for precise and reliable viscosity measurement of difficult-to-measure, highly viscous samples.
  • Fast, precise and reproducible results: The combination of ROTAVISC viscometer and ROTATEMP high-temperature system simplifies the viscosity measurement of difficult-to-measure, highly viscous samples
  • Simple operation and space-saving: The complete system is supplied ready for connection. With its space-saving design, it fits easily into any laboratory
  • Optimum compatibility and automation: ROTAVISC and
More Product Information
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR