Imprimer
Immersion Circulators, ICC 100 lite eco 8
Fournisseur: IKA
10013176
IKAA10013176EA 1530 EUR
IKAA10013176
Immersion Circulators, ICC 100 lite eco 8
Thermostats à circulation Systèmes à circulation ouverte Contrôleurs à thermostat
The compact immersion circulators of the lite series are powerful and of high quality, with a focus on essential basic functions, excellent performance and efficient operation. ICC 100 lite is suitable for tempering liquids up to 100 °C and internal circulation.
  • Max. temperature in the plastic bath (eco): 100 °C (water only)
To make configuration of your temperature control system as convenient as possible, IKA offers 'ready-to-connect' packages with all the necessary accessories.
Information de livraison: Package includes: ICC 100 lite compactMore Product Information
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR